Order Entry
Czech Republic
ContactUsLinkComponent
Molecular sieve 3A (0.3 nm, 3 Å), Hydranal™ drying agent, for air and gases for Karl Fischer titration, Ø ~2 mm, Fluka™
Molecular sieve 3A (0.3 nm, 3 Å), Hydranal™ drying agent, for air and gases for Karl Fischer titration, Ø ~2 mm, Fluka™
Catalog # 34241-250G
CAS Number:  
undefined
Molecular sieve 3A (0.3 nm, 3 Å), Hydranal™ drying agent, for air and gases for Karl Fischer titration, Ø ~2 mm, Fluka™
Catalog # 34241-250G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Suitability (for KF-titration):
    test passed

Specification Test Results

About this item

 :
Density:2 g/cm³
Storage Temperature:Ambient
CAS Number:1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613