Charcoal activated, powder, DARCO® -100 mesh, Fluka™
Katalogové číslo 242276-1KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Drum with inliner
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Particle Size:−100 mesh
- Appearance:complying
- Loss on drying (105°C):Max. 12 %
- Methylene blue adsorption:Min. 15 g/100g
- Water soluble matter:Max. 0.50 %
- In acid soluble iron (Fe):Max. 200 ppm
- Vapor Pressure:<0.1 mmHg ( 20 °C)
- Resistivity:1375 μΩ-cm, 20°C (graphite)
- Form:powder
- Autoignition Temperature:842 °F
Specification Test Results
About this item
:
:12.01 g/mol
Merck Index:13,01818
Melting Pt:3550 °C
Storage Temperature:Ambient
EINECS:231-153-3
ADR:4.2,III
REACH:01-2119488894-16
CAS Number:7440-44-0
:64365-11-3
Formula:C
MDL Number:MFCD00133992