

Hydrochloric acid fuming ≥37%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. APHA: ≤10, Fluka™
:7647-01-0
:
:HCl
:MFCD00011324
:42 °C (1.013 hPa)
:–35 °C
:1.190 g/cm³ (20 °C)
:231-595-7
:1789
:8,II
:36.46 g/mol
:GHS07
:GHS05
:Ambient
- Assay:37 - 38.0 %
- Density (20°C):Min. 1.18 g/ml
- Residue on ignition (SO₄):Max. 5 ppm
- Non-volatile matter:Max. 0.005 %
- Ammonium (NH₄):Max. 1 ppm
- Silver (Ag):Max. 0.01 ppm
- Aluminium (Al):Max. 0.000005 %
- Arsenic (As):Max. 0.01 ppm
- Barium (Ba):Max. 0.000002 %
- Beryllium (Be):Max. 0.000002 %
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.01 ppm
- Cobalt (Co):Max. 0.000001 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.2 ppm
- Germanium (Ge):Max. 0.000005 %
- Mercury (Hg):Max. 0.01 ppm
- Potassium (K):Max. 0.00001 %
- Lithium (Li):Max. 0.000001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000001 %
- Molybdenum (Mo):Max. 0.02 ppm
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.02 ppm
- Strontium (Sr):Max. 0.000001 %
- Titanium (Ti):Max. 0.00001 %
- Thallium (Tl):Max. 0.000005 %
- Vanadium (V):Max. 0.000001 %
- Zinc (Zn):Max. 0.000005 %
- Zirconium (Zr):Max. 0.00001 %
- Heavy metals (ICP) ACS:Max. 1 ppm
- Free chlorine (Cl):Max. 0.5 ppm
- Bromide (Br):Max. 0.005 %
- Phosphate (PO₄):Max. 0.00005 %
- Sulfate (SO₄):Max. 1 ppm
- Sulfite (SO₃):Max. 1 ppm
- Appearance of the substance:complying
- APHA:Max. 10
- Residual solvents:complying
Frequently Bought Together