


n-Hexan ≥99%, Reagent Grade
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD00009520
:13,04712
:66…69 °C(1,013 hPa)
:–95 °C
:0,660…0.668 g/cm³ (20 °C)
:–26 °C
:Ambient
:203-777-6
:1208
:3,II
:GHS07
:GHS08
:GHS02
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.373 - 1.377
- Vapor Pressure:256 mmHg ( 37.7 °C)
- Vapor Density:~3 (vs air)
- Refractive Index:n20/D 1.375 (lit.)
- Explosion Limit:7.7 %
Frequently Bought Together