Lithium acetate dihydrate ≥99%, Reagent Grade
Catalog # L6883-250G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:97.0 - 103.0 %
- pH (5 %, 20°C):Max. 9.5
- Water insoluble matter:Max. 0.01 %
- Iron (Fe):Max. 20 ppm
- Sodium (Na):Max. 50 ppm
- Chloride (Cl):Max. 0.003 %
- Sulfate (SO₄):Max. 100 ppm
Specification Test Results
About this item
:
:102.02 g/mol
Merck Index:13,05543
Melting Pt:53 - 56 °C
Melting Pt:53…56 °C
Density:1.3 g/cm3
Storage Temperature:Ambient
EINECS:208-914-3
CAS Number:6108-17-4
Formula:H₃CCOOLi·2H₂O
MDL Number:MFCD00066949