Order Entry
Czech Republic
ContactUsLinkComponent
Hydrobromic acid 48% ACS, Fluka™
Irritant
Corrosive
Hydrobromic acid 48% ACS, Fluka™
Certificates
Irritant
Corrosive
Hydrobromic acid 48% ACS, Fluka™

Specification Test Results

  • Vapor Pressure:
    8 mmHg ( 25 °C)
  • Assay:
    47.0 - 49.0 %
  • Residue on ignition (SO₄):
    Max. 0.002 %
  • Iron (Fe):
    Max. 1 ppm
  • Ignition Residue:
    ≤0.002%
  • Selenium (Se):
    Max. 0.01 ppm
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.05 %
  • Iodide (I):
    Max. 0.003 %
  • Phosphate (PO₄):
    Max. 0.001 %
  • Sulfate and sulfite (as SO₄):
    Max. 0.003 %

Specification Test Results

Related Information

Frequently Bought Together