Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Iron(II) sulphate heptahydrate 99.5-104.5% (by manganometry), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
 
 
Iron(II) sulphate heptahydrate 99.5-104.5% (by manganometry), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
 

 

  • Assay (manganometric):
    99.5 - 104.5 %
  • pH (5 %, 20°C):
    3.0 - 4.0
  • Arsenic (As):
    Max. 2 ppm
  • Calcium (Ca):
    Max. 0.02 %
  • Chromium (Cr):
    Max. 50 ppm
  • Copper (Cu):
    Max. 30 ppm
  • Iron(III):
    Max. 0.2 %
  • Mercury (Hg):
    Max. 1 ppm
  • Potassium (K):
    Max. 0.002 %
  • Magnesium (Mg):
    Max. 0.02 %
  • Manganese (Mn):
    Max. 0.1 %
  • Sodium (Na):
    Max. 0.01 %
  • Nickel (Ni):
    Max. 50 ppm
  • Lead (Pb):
    Max. 2 ppm
  • Zinc (Zn):
    Max. 20 ppm
  • Heavy metals (as Pb):
    Max. 0.005 %
  • Chloride (Cl):
    Max. 100 ppm
  • Residual solvents:
    complying

 

 

Frequently Bought Together