Order Entry
Czech Republic
ContactUsLinkComponent
Phenol red ACS indicator, Fluka™
Phenol red ACS indicator, Fluka™
Certificates
Phenol red ACS indicator, Fluka™
Certificates

Specification Test Results

  • Clearness of the solution:
    complying
  • Loss on drying (vacuum):
    Max. 5 %
  • Residue on ignition:
    Max. 1 %
  • Change range:
    complying
  • Extinction maximum at:
    552 - 562 nm
  • Extinction maximum of:
    900 - 1800

Specification Test Results

Related Information

Frequently Bought Together