Potassium hydroxide ≥86% (by acidometric titration), pellets, Puriss. p.a., Fluka™
Catalog # 60370-25KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Fiberboard box
- Pk:25 kg
- Seq:4283
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 86 %
- Assay of K₂CO₃:Max. 0.6 %
- Silver (Ag):Max. 0.0005 %
- Aluminium (Al):Max. 0.0005 %
- Barium (Ba):Max. 0.0005 %
- Bismuth (Bi):Max. 0.0005 %
- Calcium (Ca):Max. 0.001 %
- Cadmium (Cd):Max. 0.0005 %
- Cobalt (Co):Max. 0.0005 %
- Chromium (Cr):Max. 0.0005 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 0.0005 %
- Mercury (Hg):Max. 0.00001 %
- Lithium (Li):Max. 0.0005 %
- Magnesium (Mg):Max. 0.0005 %
- Manganese (Mn):Max. 0.0005 %
- Molybdenum (Mo):Max. 0.0005 %
- Sodium (Na):Max. 0.5 %
- Nickel (Ni):Max. 0.0005 %
- Lead (Pb):Max. 0.0005 %
- Strontium (Sr):Max. 0.0005 %
- Zinc (Zn):Max. 0.0005 %
- Chloride (Cl):Max. 0.01 %
- Phosphate (PO₄):Max. 0.0005 %
- Silicate (as SiO₂):Max. 0.0005 %
- Sulfate (SO₄):Max. 0.01 %
- Total N:Max. 0.0005 %
Specification Test Results
About this item
Synonyms: Potash caustic, Caustic potash
:
:56.11 g/mol
EINECS:215-181-3
UN:1813
REACH:01-2119487136-33
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553
Boiling Pt:1.327 °C (1.013 hPa)
Melting Pt:360 °C
Density:2.04 g/cm3 (20 °C)
ADR:8 II
Merck Index:12,07806