Silver nitrate 0.1 M AgNO3 (0.1N) in aqueous solution ≥99% Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Boiling Pt:100 °C (1.013 hPa)
Melting Pt:<0 °C
Density:1.010 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,III
:GHS05
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together