

Barium nitrate ≥99% (Ba ≥52%), Puriss. p.a. ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:10022-31-8
:
Formula:Ba(NO₃)₂
MDL Number:MFCD00003442
Merck Index:13,00986
Boiling Pt: Dec. (on heating)
Melting Pt:592 °C
Density:3.24 g/cm³ (22 °C)
Storage Temperature:Ambient
EINECS:233-020-5
ADR:5.1,II
:261.34 g/mol
UN:1446
:GHS06
:GHS03
Specification Test Results
- Assay:Min. 99.0 %
- Assay of Ba:Min. 52 %
- Water insoluble matter:Max. 0.005 %
- pH (5 %, 25 °C):5.0 - 8.0
- Calcium (Ca):Max. 0.002 %
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 2 ppm
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.002 %
- Sodium (Na):Max. 0.005 %
- Strontium (Sr):Max. 0.05 %
- Heavy metals (ICP) ACS:Max. 5 ppm
- Ammonium (NH₄):Max. 0.001 %
- Chloride (Cl):Max. 5 ppm
Specification Test Results
Frequently Bought Together