Sodium sulphate, anhydrous ≥99.0%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7757-82-6
:
Formula:Na₂SO₄
MDL Number:MFCD00003504
Merck Index:12,08829
Melting Pt:884 °C
Density:2.68 g/cm³ (20 °C)
:142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
Specification Test Results
- Assay:Min. 99.0 %
- Assay (calculated to the dried substance):99 - 101.0 %
- Loss on ignition (800°C):Max. 0.5 %
- Loss on drying (130°C):Max. 0.5 %
- Water insoluble matter:Max. 0.01 %
- pH (5 %, 25 °C):5.2 - 8.0
- Free acid (as H₂SO₄):Max. 0.05 %
- Free alkali (as NaOH):Max. 0.03 %
- Arsenic (As):Max. 0.0001 %
- Calcium (Ca):Max. 20 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 0.01 %
- Magnesium (Mg):Max. 25 ppm
- Zinc (Zn):Max. 0.02 %
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 10 ppm
- Phosphate (PO₄):Max. 0.001 %
- Total N:Max. 5 ppm
- Appearance of the solution:complying
Specification Test Results
Frequently Bought Together