Sodium sulphite ≥98.0% ACS, Fluka™
239321-500G
:
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
- Assay (Iodometric):Min. 98.0 %
- Water insoluble matter:Max. 0.005 %
- Free acid:complying
- Titrable Base:Max. 0.03 meq/g
- Iron (Fe):Max. 0.001 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 0.02 %
:
:126.04 g/mol
:13,08757
:600 °C
:2.6 g/cm³ (20 °C)
:Ambient
:231-821-4
:01-2119537420-49
:7757-83-7
:Na₂SO₃
:MFCD00003503