ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 1 L, 0.1 M NaOH (0.1 N) volumetric solution, Fluka™
Corrosive
Sodium hydroxide concentrate for dilution to 1 L, 0.1 M NaOH (0.1 N) volumetric solution, Fluka™
Certificates
Corrosive
Sodium hydroxide concentrate for dilution to 1 L, 0.1 M NaOH (0.1 N) volumetric solution, Fluka™
Specification Test Results
  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective