ContactUsLinkComponent
Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
   Sustainable Options Available
Certificates
Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
   Sustainable Options Available
Certificates
Specifications
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
  • Non-volatile matter:
    Max. 0.0003 %
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.005
  • Absorbance at 210 nm:
    Max. 0.01
  • Absorbance at 205 nm:
    Max. 0.01
  • HPLC Gradient at 254 nm:
    Max. 2 mAU
  • HPLC Gradient at 210 nm:
    Max. 10 mAU