Sodium chloride ≥99.0% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1.465 °C (1.013 hPa)
Melting Pt:800 °C
Density:2.160 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
Specification Test Results
- Assay:Min. 99.0 %
- pH (5 %, 25 °C):5.0 - 9.0
- Water insoluble matter:Max. 0.005 %
- Barium (Ba):complying
- Calcium (Ca):Max. 0.002 %
- Iron (Fe):Max. 2 ppm
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.001 %
- Heavy metals (ICP) ACS:Max. 5 ppm
- Bromide (Br):Max. 0.01 %
- Chlorate, Nitrate (as NO₃):Max. 0.003 %
- Iodide (I):Max. 0.002 %
- Phosphate (PO₄):Max. 5 ppm
- Sulfate (SO₄):Max. 0.004 %
Specification Test Results
Frequently Bought Together