Sodium hydrogen carbonate ≥99.7% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Merck Index:13,08655
Melting Pt: Dec.
Density:2.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-633-8
Specification Test Results
- Identity (IR):complying
- Assay (excluding dried substance):99.7 - 100.3 %
- Water insoluble matter:Max. 0.015 %
- Calcium (Ca):Max. 0.02 %
- Iron (Fe):Max. 0.001 %
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.005 %
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 0.003 %
- Phosphate (PO₄):Max. 0.001 %
- S-compounds (as SO₄):Max. 0.003 %
- Ammonium (NH₄):Max. 5 ppm
Specification Test Results
Frequently Bought Together