
Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:584-08-7
:
Formula:K₂CO₃
MDL Number:MFCD00011382
:138.21 g/mol
Merck Index:13,07702
Boiling Pt: Dec.
Melting Pt:891 °C
Density:2.430 g/cm³ (19 °C)
EINECS:209-529-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:99.0 - 101.0 %
- Calcium (Ca):Max. 50 ppm
- Cadmium (Cd):Max. 50 ppm
- Cobalt (Co):Max. 50 ppm
- Copper (Cu):Max. 50 ppm
- Iron (Fe):Max. 50 ppm
- Sodium (Na):Max. 5000 ppm
- Nickel (Ni):Max. 50 ppm
- Lead (Pb):Max. 50 ppm
- Zinc (Zn):Max. 50 ppm
- Total S (as SO₄):Max. 50 ppm
- Chloride (Cl):Max. 50 ppm
Specification Test Results
Frequently Bought Together