Order Entry
Czech Republic
ContactUsLinkComponent
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates
undefined
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Free acid (as HCl):
    Max. 0.001 %
  • Thiophene:
    Max. 0.0005 %
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 350 nm:
    Max. 0.01
  • Absorbance at 300 nm:
    Max. 0.10
  • Absorbance at 288 nm:
    Max. 0.50
  • Absorbance at 286 nm:
    Max. 1.00
  • Refractive Index:
    n20/D 1.496 (lit.)
  • Explosion Limit:
    7 %
  • Vapor Pressure:
    22 mmHg ( 20 °C)

Specification Test Results

Related Information

Frequently Bought Together