

Chlorobenzene ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:108-90-7
:
Formula:C₆H₅Cl
MDL Number:MFCD00000530
:112.56 g/mol
Merck Index:13,02139
Boiling Pt:130…132 °C (1.013 hPa)
Melting Pt:–45 °C
Density:1.107…1.109 g/cm³ (20 °C)
Flash Pt:28 °C
Storage Temperature:Ambient
EINECS:203-628-5
UN:1134
ADR:3,III
REACH:01-2119432722-45
:GHS02
:GHS07
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.50 %
- Titrable Acid:Max. 0.004 meq/g
- Refractive index (n 20/D):1.524 - 1.526
- Water (Karl Fischer):Max. 0.03 %
- Non-volatile matter:Max. 0.003 %
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.1 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- APHA:Max. 25
- Explosion Limit:7.1 %
- Vapor Pressure:11.8 mmHg ( 25 °C)
Specification Test Results
Frequently Bought Together