Order Entry
Czech Republic
ContactUsLinkComponent
4-(Dimethylamino)benzaldehyde ≥99.0% (by HPLC) for the determination of hydroxyproline, Fluka™
4-(Dimethylamino)benzaldehyde ≥99.0% (by HPLC) for the determination of hydroxyproline, Fluka™
Catalog # 39070-250G
CAS Number:  
undefined
4-(Dimethylamino)benzaldehyde ≥99.0% (by HPLC) for the determination of hydroxyproline, Fluka™
Catalog # 39070-250G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay (HPLC):
    Min. 99.0 %
  • Melting point:
    73 - 75 °C
  • Loss on drying:
    Max. 0.5 %
  • Colour of the solution:
    complying
  • Turbidity of solution:
    complying
  • Residue on ignition (as SO₄):
    Max. 0.1 %
  • Identity (NMR):
    complying
  • Application test:
    complying
  • Calcium (Ca):
    Max. 30 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Total S (as SO₄):
    Max. 50 ppm
  • Chloride (Cl):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :149.19 g/mol
Merck Index:13,03257
Boiling Pt:176…177 °C (23 hPa)
Melting Pt:72…75 °C
Flash Pt:164 °C
Storage Temperature:Ambient
EINECS:202-819-0
CAS Number:100-10-7
Formula:(CH₃)₂NC₆H₄CHO
MDL Number:MFCD00003381