Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl acetate ≥99.5% ACS, Riedel-de Haën™
Irritant
Flammable
Ethyl acetate ≥99.5% ACS, Riedel-de Haën™
   Sustainable Options Available
Certificates
undefined
Irritant
Flammable
Ethyl acetate ≥99.5% ACS, Riedel-de Haën™
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.50 %
  • Non-volatile matter:
    Max. 0.003 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Titrable Acid:
    Max. 0.0009 meq/g
  • Reaction against H₂SO₄:
    complying
  • APHA:
    Max. 10
  • Vapor Pressure:
    73 mmHg ( 20 °C)
  • Autoignition Temperature:
    801 °F
  • Vapor Density:
    3 (20 °C, vs air)

Specification Test Results

Related Information

Frequently Bought Together