Order Entry
Czech Republic
ContactUsLinkComponent
Disodium hydrogen phosphate dodecahydrate 98.5-101% (dried basis), meets analytical specification of BP, Ph. Eur., E339, Fluka™
Disodium hydrogen phosphate dodecahydrate 98.5-101% (dried basis), meets analytical specification of BP, Ph. Eur., E339, Fluka™
Dodavatel:  Honeywell Chemicals
Certifikáty
Disodium hydrogen phosphate dodecahydrate 98.5-101% (dried basis), meets analytical specification of BP, Ph. Eur., E339, Fluka™
Dodavatel:  Honeywell Chemicals
Certifikáty

Výsledky testů specifikací

  • Assay:
    98.5 - 102.5 %
  • Assay (calculated to the dried substance):
    98.5 - 101 %
  • Assay of NaH₂PO₄ (calculated to dried substances):
    Max. 1.0 %
  • Water (Karl Fischer):
    57.0 - 61.0 %
  • Loss on drying (130°C):
    Max. 61.0 %
  • Water insoluble matter:
    Max. 0.2 %
  • pH (1 %,20°C):
    8.4 - 9.6
  • Arsenic (As):
    Max. 1 ppm
  • Cadmium (Cd):
    Max. 1 ppm
  • Copper (Cu):
    Max. 0.002 %
  • Iron (Fe):
    Max. 10 ppm
  • Mercury (Hg):
    Max. 1 ppm
  • Potassium (K):
    Max. 0.005 %
  • Lead (Pb):
    Max. 1 ppm
  • Zinc (Zn):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Chloride (Cl):
    Max. 10 ppm
  • Fluoride (F):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 100 ppm
  • Appearance of the solution:
    complying
  • Reducing impurities:
    complying
  • Residual solvents:
    complying

Výsledky testů specifikací

Související informace

Frequently Bought Together