Order Entry
Czech Republic
ContactUsLinkComponent
 
Human Recombinant IL-31 (from E. coli)
 
undefined
Human Recombinant IL-31 (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: STAT3 Activation in U-87 MG cells, ED50 ≤Detectable at ≤ 25 ng/ml, bioactivity ≥0
  • Gene ID:
    Q6EBC2
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    IL-31|Interleukin-31
  • Protein/peptide name:
    IL31
  • Purity:
    ≥95%
  • Molecular weight:
    15,8 kDa
  • Sequence:
    SHTLPVRLLR PSDDVQKIVE ELQSLSKMLL KDVEEEKGVL VSQNYTLPCL SPDAQPPNNI HSPAIRAYLK TIRQLDNKSV IDEIIEHLDK LIFQDAPETN ISVPTDTHEC KRFILTISQQ FSECMDLALK SLTSGAQQAT T
  • Endotoxin level:
    Low
  • Formulation:
    20 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together