Order Entry
Czech Republic
ContactUsLinkComponent
 
Human Recombinant BMP-4 (from CHO cells)
 
undefined
Human Recombinant BMP-4 (from CHO cells)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: Alkaline phosphatase activity induced in ATDC-5 cells, ED50 ≤15 ng/ml, bioactivity ≥6,7×10⁴ units/mg
  • Gene ID:
    P12644
  • Reconstitution instructions:
    Sterile 10 mM HCl at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    BMP-2B
  • Protein/peptide name:
    BMP4
  • Purity:
    ≥95%
  • Molecular weight:
    13,1/26,2 kDa
  • Sequence:
    SPKHHSQRAR KKNKNCRRHS LYVDFSDVGW NDWIVAPPGY QAFYCHGDCP FPLADHLNST NHAI VQT LVN SVNSSIPKAC CVPTELSAIS MLYLDEYDKV VLKNYQEMVV EGCGCR
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together