ContactUsLinkComponent
J.T.Baker®, Cycle-Tap Sampler
J.T.Baker®, Cycle-Tap Sampler
Catalog # 9815-05
Supplier:  Avantor
undefined
J.T.Baker®, Cycle-Tap Sampler
Catalog # 9815-05
Supplier:  Avantor
Specifications
  • Colour:
    Amber
  • Description:
    Cycle-tap sampler
  • Seq:
    05
  • Pk:
    1
Specifications
About this item
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.