Zadání objednávky
Czech Republic
ContactUsLinkComponent
Dextran, M.W. 60000 - 90000, low fraction, J.T.Baker®
Dextran, M.W. 60000 - 90000, low fraction, J.T.Baker®
Catalog # G200-05
Supplier:  Avantor
CAS Number:  
undefined
Dextran, M.W. 60000 - 90000, low fraction, J.T.Baker®
Catalog # G200-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle, amber, wide mouth, black T/E cap
  • Bal.:
    100 g
  • Sekvence:
    3401

Specifications

Specification Test Results

  • Appearance:
    Passes Test

Specification Test Results

About this item

 :
CAS Number:9004-54-0
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935