Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Amisulbrom, reference material, Honeywell Fluka™
  HONCR1360-25MG
 :  
Analytical standard, Amisulbrom, reference material, Honeywell Fluka™
  HONCR1360-25MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    348635-87-0
  • Description:
    Amisulbrom
  • Melting point:
    127…132 °C
  • Formula:
    C₁₃H₁₃BrFN₅O₄S₂
  • Storage temperature:
    Ambient
  • Description:
    Amisulbrom

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results