Analytical standard, Monosultap, reference material, Honeywell Fluka™
Catalog # HONCR1359-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard, neat
- CAS No.:29547-00-0
- Description:Monosultap
- Melting point:142…143 °C
- Formula:C₅H₁₂NNaO₆S₄
- Storage temperature:Ambient
- Description:Monosultap
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results