Order Entry
Czech Republic
ContactUsLinkComponent
Analytical standard, Melamine-13C3, reference material, Honeywell Fluka™
Analytical standard, Melamine-13C3, reference material, Honeywell Fluka™
Catalog # HONCR1355-10MG
CAS Number:  
Analytical standard, Melamine-13C3, reference material, Honeywell Fluka™
Catalog # HONCR1355-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1173022-88-2
  • Description:
    Melamine-13C3
  • Melting point:
    min. 300 °C
  • Formula:
    ¹³C₃H₆N₆
  • Storage temperature:
    Ambient
  • Description:
    Melamine-13C3

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results