Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Mapenterol-(dimethyl-d₆, propyl-d₅) hydrochloride, reference material, Honeywell Fluka™
  HONCR1342-10MG
 :  
Analytical standard, Mapenterol-(dimethyl-d₆, propyl-d₅) hydrochloride, reference material, Honeywell Fluka™
  HONCR1342-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1325559-18-9
  • Description:
    Mapenterol-(dimethyl-d₆, propyl-d₅) hydrochloride
  • Melting point:
    177…179 °C
  • Formula:
    C₁₄D₁₁H₉ClF₃N₂O · HCl
  • Storage temperature:
    Ambient
  • Description:
    Mapenterol-(dimethyl-d₆, propyl-d₅) hydrochloride

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results