Analytical standard, Ochratoxin A-(phenyl-d₅) solution reference material, Honeywell Fluka™
Katalogové číslo HONCR1324-2ML
Dodavatel: Honeywell Chemicals
Číslo CAS:
Specifikace
- Concentration:10 μg/ml in acetonitrile
- Pack type:Glass Bottle
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:666236-28-8
- Description:Ochratoxin A-(phenyl-d₅) solution
- Melting point:–45 °C
- Formula:C₂₀D₅H₁₃ClNO₆
- Storage temperature:–20°C
- Description:Ochratoxin A-(phenyl-d₅) solution
- Boiling point:82 °C
Specifikace
O tomto produktu
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results