Analytical standard, Zearalanone solution, reference material, Honeywell Fluka™
Catalog # HONCR1322-5MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:5 mg
- Standards:Analytical standard, neat
- CAS No.:5975-78-0
- Description:Zearalanone solution
- Melting point:190…192 °C
- Formula:C₁₈H₂₄O₅
- Storage temperature:–20°C
- Description:Zearalanone solution
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results