Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Prochloraz Metabolite BTS40348, reference material, Honeywell Fluka™
  HONCR1290-10MG
 :  
Analytical standard, Prochloraz Metabolite BTS40348, reference material, Honeywell Fluka™
  HONCR1290-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    67747-01-7
  • Description:
    Prochloraz Metabolite BTS40348
  • Formula:
    C₁₁H₁₄Cl₃NO
  • Storage temperature:
    Ambient
  • Description:
    Prochloraz Metabolite BTS40348

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results