Analytical standard, (±)-3-Chloro-1,2-propanediol, reference material, Honeywell Fluka™
HONCR1287-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard, neat
- CAS No.:96-24-2
- Description:(±)-3-Chloro-1,2-propanediol
- Melting point:–52…–40 °C
- Formula:ClCH₂CH(OH)CH₂OH
- Storage temperature:Ambient
- Description:(±)-3-Chloro-1,2-propanediol
- Boiling point:216…219 °C
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results