Analytical standard, Imidacloprid-olefin, reference material, Honeywell Fluka™
Catalog # HONCR1278-10MG
Dodavatel: Honeywell Chemicals
Číslo CAS:
Specifikace
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:115086-54-9
- Description:Imidacloprid-olefin
- Melting point:187…189 °C
- Formula:C₉H₈ClN₅O₂
- Storage temperature:2…8 °C
- Description:Imidacloprid-olefin
Specifikace
O tomto produktu
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results