Analytical standard, Toltrazuril-(N-methyl-d₃), reference material, Honeywell Fluka™
Katalogové číslo HONCR1247-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1353867-75-0
- Description:Toltrazuril-(N-methyl-d₃)
- Melting point:191…197 °C
- Formula:C₁₈D₃H₁₁F₃N₃O₄S
- Storage temperature:Ambient
- Description:Toltrazuril-(N-methyl-d₃)
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results