Order Entry
Czech Republic
ContactUsLinkComponent
Analytical standard, Sulfadimethoxine-(phenyl-¹³C₆), reference material, Honeywell Fluka™
Analytical standard, Sulfadimethoxine-(phenyl-¹³C₆), reference material, Honeywell Fluka™
Catalog # HONCR1244-10MG
CAS Number:  
Analytical standard, Sulfadimethoxine-(phenyl-¹³C₆), reference material, Honeywell Fluka™
Catalog # HONCR1244-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1334378-48-1
  • Description:
    Sulfadimethoxine-(phenyl-¹³C₆)
  • Melting point:
    203 °C
  • Formula:
    ¹³C₆C₆H₁₄N₄O₄S
  • Storage temperature:
    Ambient
  • Description:
    Sulfadimethoxine-(phenyl-¹³C₆)

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results