Analytical standard, Sulfaguanidin, reference material, Honeywell Fluka™
HONCR1243-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:227622-73-3
- Description:Sulfaguanidin
- Melting point:75…80 °C
- Formula:C₇H₁₁N₃O₃ · 0.5H₂O
- Storage temperature:Ambient
- Description:Sulfaguanidin
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results