Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Mabuterol-(tert-butyl-d₉) hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1238-10MG
CAS Number:  
Analytical standard, Mabuterol-(tert-butyl-d₉) hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1238-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1353867-83-0
  • Description:
    Mabuterol-(tert-butyl-d₉) hydrochloride
  • Melting point:
    202…204 °C
  • Formula:
    C₁₃D₉H₉ClF₃N₂O · HCl
  • Storage temperature:
    Ambient
  • Description:
    Mabuterol-(tert-butyl-d₉) hydrochloride

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results