Order Entry
Czech Republic
ContactUsLinkComponent
Analytical standard, Piroxicam-(methyl-d₃), reference material, Honeywell Fluka™
Analytical standard, Piroxicam-(methyl-d₃), reference material, Honeywell Fluka™
Katalogové číslo HONCR1205-10MG
Dodavatel:  Honeywell Chemicals
Číslo CAS:  
Analytical standard, Piroxicam-(methyl-d₃), reference material, Honeywell Fluka™
Katalogové číslo HONCR1205-10MG
Dodavatel:  Honeywell Chemicals
Číslo CAS:  

Specifikace

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    942047-64-5
  • Description:
    Piroxicam-(methyl-d₃)
  • Melting point:
    198…200 °C
  • Formula:
    C₁₅D₃H₁₀N₃O₄S
  • Storage temperature:
    Ambient
  • Description:
    Piroxicam-(methyl-d₃)

Specifikace

O tomto produktu

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results