Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Rafoxanide-(benzoyl ring-¹³C₆), reference material, Honeywell Fluka™
Catalog # HONCR1198-10MG
CAS Number:  
Analytical standard, Rafoxanide-(benzoyl ring-¹³C₆), reference material, Honeywell Fluka™
Catalog # HONCR1198-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1353867-98-7
  • Description:
    Rafoxanide-(benzoyl ring-¹³C₆)
  • Melting point:
    145…150 °C
  • Formula:
    ¹³C₆C₁₃H₁₁Cl₂I₂NO₃
  • Storage temperature:
    Ambient
  • Description:
    Rafoxanide-(benzoyl ring-¹³C₆)

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results