Analytical standard, Nafcillin sodium salt, reference material, Honeywell Fluka™
Catalog # HONCR1185-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard, neat
- CAS No.:985-16-0
- Description:Nafcillin sodium salt
- Melting point:168…288 °C
- Formula:C₂₁H₂₁N₂NaO₅S
- Storage temperature:–20°C
- Description:Nafcillin sodium salt
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results