Analytical standard, Decoquinate, reference material, Honeywell Fluka™
Catalog # HONCR1176-50MG
Dodavatel: Honeywell Chemicals
Číslo CAS:
Specifikace
- Pack type:Glass Bottle
- Pk:50 mg
- Standards:Analytical standard, neat
- CAS No.:18507-89-6
- Description:Decoquinate
- Melting point:242…246 °C
- Formula:C₂₄H₃₅NO₅
- Storage temperature:Ambient
- Description:Decoquinate
Specifikace
O tomto produktu
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results