Analytical standard, Metoclopramide, reference material, Honeywell Fluka™
Catalog # HONCR1172-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard, neat
- CAS No.:364-62-5
- Description:Metoclopramide
- Environmentally Preferable:
- Melting point:147 °C
- Formula:C₁₄H₂₂ClN₃O₂
- Storage temperature:2…8 °C
- Description:Metoclopramide
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results