Analytical standard, Zilpaterol-(isopropyl-13C3) hydrochloride, reference material, Honeywell Fluka™
HONCR1158-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1613439-55-6
- Description:Zilpaterol-(isopropyl-13C3) hydrochloride
- Melting point:228 °C
- Formula:¹³C₃C₁₁H₁₉N₃O₂ · HCl
- Storage temperature:2…8 °C
- Description:Zilpaterol-(isopropyl-13C3) hydrochloride
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results