Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Tralopyril, reference material, Honeywell Fluka™
  HONCR1105-100MG
 :  
Analytical standard, Tralopyril, reference material, Honeywell Fluka™
  HONCR1105-100MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    122454-29-9
  • Description:
    Tralopyril
  • Melting point:
    252…254 °C
  • Formula:
    C₁₂H₅BrClF₃N₂
  • Storage temperature:
    Ambient
  • Description:
    Tralopyril

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results