Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Detomidine hydrochloride monohydrate, reference material, Honeywell Fluka™
Catalog # HONCR1056-10MG
CAS Number:  
Analytical standard, Detomidine hydrochloride monohydrate, reference material, Honeywell Fluka™
Catalog # HONCR1056-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    90038-00-9
  • Description:
    Detomidine hydrochloride monohydrate
  • Melting point:
    157…159 °C
  • Formula:
    C₁₂H₁₄N₂ · HCl · H₂O
  • Storage temperature:
    Ambient
  • Description:
    Detomidine hydrochloride monohydrate

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results