Analytical standard, Secbumeton-ethyl, reference material, Honeywell Fluka™
Catalog # HONCR1045-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1705649-53-1
- Description:Secbumeton-ethyl
- Formula:C₁₀D₅H₁₄N₅O
- Storage temperature:Ambient
- Description:Secbumeton-ethyl
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results