Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Azaperon-d₄, reference material, Honeywell Fluka™
  HONCR1026-10MG
 :  
Analytical standard, Azaperon-d₄, reference material, Honeywell Fluka™
  HONCR1026-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1173021-72-1
  • Description:
    Azaperon-d₄
  • Melting point:
    91…92 °C
  • Formula:
    C₁₉D₄H₁₈FN₃O
  • Storage temperature:
    Ambient
  • Description:
    Azaperon-d₄

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results